Drukuj
Łapy trójpalczaste z korkiem
Dostawca: USBECK Laborgeräte
2800 2801 2802
241-7431EA 55 PLN
241-7431 241-7432 241-7433
Łapy trójpalczaste z korkiem
Zaciski Zaciski uniwersalne
Zinc die-cast, powder coated.
  • The prongs of the clamps are covered with a plastic coating to protect the items clamped
  • Without bosshead for fixing to stand
  • With wing screws for rapid easy fixing