Drukuj
Pipety gruszkowe, dwa znaki, klasa AS
Dostawca: Bohlender
H972-02 H972-04
459-9125EA 414 PLN
459-9125 534-0000
Pipety gruszkowe, dwa znaki, klasa AS
Kamyczki wrzenne
Boiling stones, made of PTFE, white, prevent chemical splashes and production of big bubbles when boiling the medium. Grain size: 4 mm. Packed in resealable bag. FDA conformity.
  • Temperature resistance: From −200 to +250 °C
  • Durable
  • Excellent chemical resistance