Drukuj
Film do obrazowania chemiluminescencyjnego, Kodak BioMax® Light
Dostawca: CARESTREAM HEALTH
1538966 1917012 8689358 8761520
730-3211EA 0 PLN
730-3211 730-3226 730-3238 730-3244
Film do obrazowania chemiluminescencyjnego, Kodak BioMax® Light
Filmy do autoradiografii Filmy
BioMax® Light is a double emulsion film that provides maximum clarity and sensitivity for detection of chemiluminescent-labelled samples. BioMax® Light produces the highest signal-to-noise ratio of any Kodak film for detection of chemiluminescence. This film performs well with all brands of chemiluminescent kits. BioMax® Light film is suitable for direct and intensifying screen exposures with commonly used isotopes, though it provides lower sensitivity resulting in longer exposure times compared to BioMax® MS, and X-OMAT AR films for radioisotope detection.
  • Suitable for both manual and automated processing
Darkroom handling: BioMax® Light film should be handled under safelight conditions using a safelight lamp equipped with a Safelight Filter GBX-2 and 15 W frosted bulb (other safelight filters may result in increased background fog and/or unwanted image exposure). The safelight should be positioned 120 cm from the film at all times. Minimise the length of time that the film is exposed to safelight conditions.