Drukuj
Lupy do czytania, powerlux
Dostawca: ESCHENBACH OPTIK
158620 158628
630-1223EA 953 PLN
630-1223 630-2182
Lupy do czytania, powerlux
Lupy
These illuminated magnifiers have a Ceratec® anti-scratch hard coating ensuring reliability, comfort and a relaxed vision.
  • Comfortable use
  • Automatic switch off after 30 minutes
  • Compact design for intuitive movement on the material
They consistently provide bright and glare-free illumination thanks to two SMD LEDs (8000 K).
Dodatkowe informacje: Supplied with a protective hard foam case and three AA batteries.