Drukuj
PurFlock® DRYSWAB™ Flocked Specimen Swabs
Dostawca: MEDICAL WIRE EQUIPMENT
MW810 MW813 MW833
MEDWMW810EA 4510 PLN
MEDWMW810 MEDWMW813 MEDWMW833
PurFlock® DRYSWAB™ Flocked Specimen Swabs
Wymazówki Wymazówki do próbkowania
Flocked swabs are ideal for PCR, molecular assays, forensic applications, food applications, and direct antigen testing. The bud is formed by a layer of tiny multi-filament polyester fibres of varying lengths, increasing the effective surface area of the swab.
  • Higher absorption and release capacity in comparison to standard flock swabs
  • Multi-length fibers that are ultrafine
  • Proprietary adhesive
  • Unique micro-structure of fibers
  • Rapid absorption and release
  • Quick and easy release of specimen
  • Flexible
Used for recovery of bacterial or viral organisms, rapid diagnostic testing, environmental sampling, food safety testing, forensics, buccal cell collection, DNA testing, nasopharyngeal or urethral specimen collection.
Shelf life: 6 months.
Certyfikaty: ISO 13485: 2016, CLSI M40-A2, FDA GMP’s.