Drukuj

Vanadyl sulphate hydrate, Sigma-Aldrich®

Dostawca: Merck
Uwaga

Synonim: Vanadium (IV) sulphate oxide hydrate, Vanadium(IV) oxide sulphate hydrate, Vanadium oxide sulphate hydrate, Basic vanadium (IV) sulphate hydrate

M-Clarity™ quality level = MQ200

204862-10G 204862-50G
SIAL204862-10GEA 609 PLN
SIAL204862-10G SIAL204862-50G
Vanadyl sulphate hydrate, Sigma-Aldrich®
Vanadyl sulphate hydrate
Formuła: O₅SV·H₂O
MW: 163.00 g/mol (anhy)
Numer MDL: MFCD00149933
Numer CAS: 123334-20-3
Numer WE (dawny EINECS): 248-652-7
Jednostka: 2931
ADR: 6.1 II

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR