Drukuj
Celulaza Trichoderma reesei
Dostawca: Merck
C8546-10KU C8546-2.5KU C8546-5KU
SIALC8546-10KUEA 820 PLN
SIALC8546-10KU SIALC8546-2.5KU SIALC8546-5KU
Celulaza Trichoderma reesei
Enzymy
Cellulase from Sigma has been used to degrade cello-oligosaccharides into glucose; to study the biodegradability of bioabsorbable bacterial cellulose (BBC) material. Cellulase from Trichoderma reesei has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. Cellulase from Trichoderma reesei has also been used in a study to investigate the liquefaction of hydrothermally pretreated wheat straw at high-solids content.