Drukuj

Anti-Gastric Mucin Mouse Monoclonal Antibody [clone: ABT-MUC6]

Dostawca: ANTIBODIES.COM
A100017-100 A100017-50
ANTIA100017-100EA 2380 PLN
ANTIA100017-100 ANTIA100017-50
Anti-Gastric Mucin Mouse Monoclonal Antibody [clone: ABT-MUC6]
Przeciwciała
Mouse monoclonal (ABT-MUC6) antibody to MUC6 for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: Gastric Mucin
Clonality: Monoclonal
Clone: ABT-MUC6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR