Drukuj

Anti-CD1a Mouse Monoclonal Antibody [clone: ABT-CD1a]

Dostawca: ANTIBODIES.COM
A100036-100 A100036-50
ANTIA100036-100EA 2380 PLN
ANTIA100036-100 ANTIA100036-50
Anti-CD1a Mouse Monoclonal Antibody [clone: ABT-CD1a]
Przeciwciała
Mouse monoclonal (ABT-CD1a) antibody to CD1a for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: ABT-CD1a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR