Drukuj
Anti-CD45RO Mouse Monoclonal Antibody [clone: ABT-CD45]
Dostawca: ANTIBODIES.COM
A100028-100 A100028-50
ANTIA100028-100EA 2380 PLN
ANTIA100028-100 ANTIA100028-50
Anti-CD45RO Mouse Monoclonal Antibody [clone: ABT-CD45]
Przeciwciała
Mouse monoclonal (ABT-CD45) antibody to CD45 for IHC with samples derived from human.
  • Validated Applications: IHC
Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: ABT-CD45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human