Drukuj
Anti-IL3RA / CD123 Rabbit Recombinant Antibody [clone: DM33] (Biotin)
Dostawca: ANTIBODIES.COM
A317424-100 A317424-500
ANTIA317424-100EA 4630 PLN
ANTIA317424-100 ANTIA317424-500
Anti-IL3RA / CD123 Rabbit Recombinant Antibody [clone: DM33] (Biotin)
Przeciwciała
Recombinant rabbit monoclonal [DM33] antibody to IL3RA / CD123 (Biotin) for ELISA and Flow Cytometry with samples derived from human.
  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: IL3RA / CD123
Clonality: Recombinant
Clone: DM33
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human