Drukuj

Anti-Calretinin Mouse Monoclonal Antibody [clone: ABT-Calret 1]

Dostawca: ANTIBODIES.COM
A96687-100 A96687-50
ANTIA96687-100EA 2380 PLN
ANTIA96687-100 ANTIA96687-50
Anti-Calretinin Mouse Monoclonal Antibody [clone: ABT-Calret 1]
Przeciwciała
Mouse monoclonal (ABT-Calret 1) antibody to Calretinin for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Calretinin
Clonality: Monoclonal
Clone: ABT-Calret 1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR