Drukuj

Anti-Cytokeratin 5 + 6 Mouse Monoclonal Antibody [clone: ABT-CK5/6]

Dostawca: ANTIBODIES.COM
A95686-100 A95686-50
ANTIA95686-100EA 2380 PLN
ANTIA95686-100 ANTIA95686-50
Anti-Cytokeratin 5 + 6 Mouse Monoclonal Antibody [clone: ABT-CK5/6]
Przeciwciała
Mouse monoclonal (ABT-CK5/6) antibody to Cytokeratin 5 + 6 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: ABT-CK5/6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR