Drukuj
Anti-Nestin Mouse Monoclonal Antibody [clone: 4D11]
Dostawca: ANTIBODIES.COM
A85293-100 A85293-50
ANTIA85293-100EA 2600 PLN
ANTIA85293-100 ANTIA85293-50
Anti-Nestin Mouse Monoclonal Antibody [clone: 4D11]
Przeciwciała
Mouse monoclonal (4D11) antibody to Nestin for WB, ICC/IF and IHC with samples derived from human, rat and mouse.
  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:500, ICC/IF: 1:50
Type: Primary
Antigen: Nestin
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse