Drukuj

Anti-Glucose Transporter GLUT1 Mouse Monoclonal Antibody [clone: ABT-GLUT1]

Dostawca: ANTIBODIES.COM
A99326-100 A99326-50
ANTIA99326-100EA 2380 PLN
ANTIA99326-100 ANTIA99326-50
Anti-Glucose Transporter GLUT1 Mouse Monoclonal Antibody [clone: ABT-GLUT1]
Przeciwciała
Mouse monoclonal (ABT-GLUT1) antibody to GLUT 1 for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Glucose Transporter GLUT1
Clonality: Monoclonal
Clone: ABT-GLUT1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR