Drukuj
Anti-Gastrin I Mouse Monoclonal Antibody [clone: ABT-GAST]
Dostawca: ANTIBODIES.COM
A99260-100 A99260-50
ANTIA99260-100EA 2380 PLN
ANTIA99260-100 ANTIA99260-50
Anti-Gastrin I Mouse Monoclonal Antibody [clone: ABT-GAST]
Przeciwciała
Mouse monoclonal (ABT-GAST) antibody to Gastrin for IHC with samples derived from human.
  • Validated applications: IHC
Type: Primary
Antigen: Gastrin I
Clonality: Monoclonal
Clone: ABT-GAST
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human