Drukuj

Anti-Glucagon Mouse Monoclonal Antibody [clone: ABT-GCG]

Dostawca: ANTIBODIES.COM
A99249-100 A99249-50
ANTIA99249-100EA 2380 PLN
ANTIA99249-100 ANTIA99249-50
Anti-Glucagon Mouse Monoclonal Antibody [clone: ABT-GCG]
Przeciwciała
Mouse monoclonal (ABT-GCG) antibody to Glucagon for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Glucagon
Clonality: Monoclonal
Clone: ABT-GCG
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR