Drukuj

Anti-CD31 Mouse Monoclonal Antibody [clone: ABT-CD31]

Dostawca: ANTIBODIES.COM
A99232-100 A99232-50
ANTIA99232-100EA 2380 PLN
ANTIA99232-100 ANTIA99232-50
Anti-CD31 Mouse Monoclonal Antibody [clone: ABT-CD31]
Przeciwciała
Mouse monoclonal (ABT-CD31) antibody to CD31 for IHC and WB with samples derived from human.

  • Validated applications: IHC, WB

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: ABT-CD31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR