Drukuj

Anti-Caldesmon Mouse Monoclonal Antibody [clone: ABT-CALD]

Dostawca: ANTIBODIES.COM
A97075-100 A97075-50
ANTIA97075-100EA 2380 PLN
ANTIA97075-100 ANTIA97075-50
Anti-Caldesmon Mouse Monoclonal Antibody [clone: ABT-CALD]
Przeciwciała
Mouse monoclonal (ABT-CALD) antibody to Caldesmon for IHC and WB with samples derived from human.

  • Validated applications: IHC, WB

Type: Primary
Antigen: Caldesmon
Clonality: Monoclonal
Clone: ABT-CALD
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR