Drukuj

Anti-IL-17RA Receptor Rabbit Recombinant Antibody [clone: DM125]

Dostawca: ANTIBODIES.COM
A318582-100 A318582-500
ANTIA318582-100EA 3200 PLN
ANTIA318582-100 ANTIA318582-500
Anti-IL-17RA Receptor Rabbit Recombinant Antibody [clone: DM125]
Przeciwciała
Recombinant rabbit monoclonal [DM125] antibody to IL-17RA Receptor for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: IL-17RA Receptor
Clonality: Recombinant
Clone: DM125
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR