Drukuj

Anti-IgG Goat Polyclonal Antibody (mFluor™ Red 780)

Dostawca: AAT BIOQUEST
49580 49581
AATB49580EA 1250 PLN
AATB49580 AATB49581
Anti-IgG Goat Polyclonal Antibody (mFluor™ Red 780)
Przeciwciała
mFluor™ Red 780 goat anti-rabbit conjugates are secondary antibodies designed for optimal performance in immunoassay applications, including flow cytometry, immunofluorescence, and confocal microscopy.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

mFluor™ Red 780 is optimally excited by the red laser and emits NIR fluorescence maximally at 767 nm.

Uwaga: Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: mFluor™ Red 780
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR