Drukuj
Anti-GPCR RDC1/CXCR-7 Recombinant Recombinant Antibody [clone: DMC443]
Dostawca: ANTIBODIES.COM
A318735-100 A318735-500
ANTIA318735-100EA 3200 PLN
ANTIA318735-100 ANTIA318735-500
Anti-GPCR RDC1/CXCR-7 Recombinant Recombinant Antibody [clone: DMC443]
Przeciwciała
Recombinant chimeric monoclonal [DMC443] antibody to GPCR RDC1/CXCR-7 for flow cytometry with samples derived from human.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: GPCR RDC1/CXCR-7
Clonality: Recombinant
Clone: DMC443
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human