Drukuj

Anti-Desmin Mouse Monoclonal Antibody [clone: ABT-DES]

Dostawca: ANTIBODIES.COM
A99759-100 A99759-50
ANTIA99759-100EA 2380 PLN
ANTIA99759-100 ANTIA99759-50
Anti-Desmin Mouse Monoclonal Antibody [clone: ABT-DES]
Przeciwciała
Mouse monoclonal (ABT-DES) antibody to Desmin for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: Desmin
Clonality: Monoclonal
Clone: ABT-DES
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR