Drukuj

Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 514)

Dostawca: AAT BIOQUEST
50062 50063
AATB50062EA 717 PLN
AATB50062 AATB50063
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 514)
Przeciwciała
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

Uwaga: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 514
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR