Drukuj
Cryogenic Thermal Transfer Labels for Frozen Surfaces
Dostawca: GA International
GAINL2FS-5C1-1WHEA 620 PLN
GAINL2FS-5C1-1WH GAINL2FS327C32.5WH GAINL2FS-3C1-1WH GAINL2FS6NPNOT34WH GAINL2FS-4C3-2WH GAINL2FS6NPNOT11WH GAINL2FS312NOT11WH GAINL2FS312NOT33WH GAINL2FS-2C3-4WH GAINL2FS6NOTC1-1WH GAINL2FS-3C1.5-1WH GAINL2FS-10C3-4WH GAINL2FS327NOT11WH GAINL2FS-7C3-2WH GAINL2FS345NOT33WH GAINL2FS345NOT11WH GAINL2FS-5C3-1WH GAINL2FS-4C1-2WH GAINL2FS308NOT32WH GAINL2FS308NOT11WH GAINL2FS-12C3-2WH GAINL2FS-12C1-1WH GAINL2FS-3C3-1WH GAINL2FS-2C1-2WH GAINL2FS-7C1-1WH GAINL2FS-10C1-2WH GAINL2FS6NOTC3-4WH GAINL2FS-1C3-5WH
Cryogenic Thermal Transfer Labels for Frozen Surfaces
Etykiety
Thermal transfer cryogenic labels that will rapidly adhere to already frozen surfaces under deep-freeze conditions −80 °C. Designed for long-term preservation in liquid nitrogen −196 °C and ultra-low temperature lab freezers, these labels will also withstand temperatures up to 100° C, perfect for use in a variety of lab applications.
  • Temperature range: −196 to +100 °C
  • Adhere quickly to already frozen surfaces (no thawing required)
  • Ideal for long-term storage in cryogenic conditions –196° C
  • Extra-permanent adhesive, ensures strong adhesion even to wet surfaces
  • Withstand temperatures up to 100 °C
Cryogenic labels, with a clear overlap area, designed to be applied directly, and rapidly adhere to frozen plastic and glass cryo vials and tubes which are difficult to label under deep-freeze conditions −80 °C. Designed for long-term preservation in liquid nitrogen −196 °C, −80 °C freezers, and dry ice, and can withstand temperatures up to 100 °C. Labels can be printed with all major brands of thermal transfer printers, or written on with cryo markers. It is recommended that the labels are applied with an overlap of at least 6,35 mm to ensure proper adherence to the frozen surface.