Drukuj
Cohesive Pocket Samplers
Dostawca: Sampling Systems
2220B-600 2220B-1500 2220B-900
SAMP2220B-600EA 9980 PLN
SAMP2220B-600 SAMP2220B-1500 SAMP2220B-900
Cohesive Pocket Samplers
Próbniki Próbniki do ciał stałych
The cohesive pocket sampler is designed for taking discreet samples of cohesive powders.
  • Ingenious scraper pushes powder into containment when rotated
  • Made from 316L stainless steel
  • Available from stock
  • Fast delivery
The cohesive pocket sampler is perfect for taking discreet samples of powders with poor flow characteristics. The scraper pushes powder into the sampler when rotated, while a solid inner rod ensures that samples are kept separate from each other.