Drukuj
ESD Safety Shoes, Lacing System, Matthew Pro Boa® GTX Low S3 CI Type 2
Dostawca: ELTEN
1728502/36 1728502/37 1728502/38 1728502/39 7285502/40 7285502/47 7285502/48 7285502/41 7285502/42 7285502/43 7285502/44 7285502/45 7285502/46
ELTN1728502/36EA 724 PLN
ELTN1728502/36 ELTN1728502/37 ELTN1728502/38 ELTN1728502/39 ELTN7285502/40 ELTN7285502/47 ELTN7285502/48 ELTN7285502/41 ELTN7285502/42 ELTN7285502/43 ELTN7285502/44 ELTN7285502/45 ELTN7285502/46
ESD Safety Shoes, Lacing System, Matthew Pro Boa® GTX Low S3 CI Type 2
Obuwie i buty
ESD safety shoe made from hydrophobised microfibre and hydrophobised textile Cordura® upper and a PU/PU sole (Ergo-Active 2). Type 2: For average wide feet with long toes, medium ball and heel width, flat ball angle.
  • Gore-Tex Crosstech® membrane
  • Full, padded bellows tongue
  • Boa® fit lacing system
  • Semi-orthopaedic inlay sole ESD black
  • Non-metallic, penetration-resistant midsole
  • Steel toe cap
  • TPU scuff cap
Suitable for indoors and outdoors, areas with exposure to moisture (S2), risk of electrostatic discharge (ESDS/ESD) and areas where there is risk of penetration from pointed and sharp objects (S3).
Colour: Black
Shoe size for ladies: 36 to 39
Shoe size for men: 40 to 48
Certyfikaty: EN ISO 20345 S3 CI/SRC, form A.