Drukuj
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Blue 570)
Dostawca: AAT BIOQUEST
103800T0 103800T1
AATB103800T0EA 1740 PLN
AATB103800T0 AATB103800T1
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Blue 570)
Przeciwciała
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Uwaga: For research use only.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human