Drukuj

Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC (Allophycocyanin))

Dostawca: AAT BIOQUEST
100331C0 100331C1 100331C2
AATB100331C0EA 1270 PLN
AATB100331C0 AATB100331C1 AATB100331C2
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC (Allophycocyanin))
Przeciwciała
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR