Drukuj
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PerCP)
Dostawca: AAT BIOQUEST
101911T0 101911T1 101911T2
AATB101911T0EA 1270 PLN
AATB101911T0 AATB101911T1 AATB101911T2
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PerCP)
Przeciwciała
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Uwaga: For research use only.
Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype:
Reactivity: Human