Drukuj
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/iFluor® 750)
Dostawca: AAT BIOQUEST
101911R0 101911R1 101911R2
AATB101911R0EA 1500 PLN
AATB101911R0 AATB101911R1 AATB101911R2
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/iFluor® 750)
Przeciwciała
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Uwaga: For research use only.
Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PE/iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human