Drukuj

Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/iFluor® 594)

Dostawca: AAT BIOQUEST
101911Y0 101911Y1 101911Y2
AATB101911Y0EA 1500 PLN
AATB101911Y0 AATB101911Y1 AATB101911Y2
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/iFluor® 594)
Przeciwciała
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR