Drukuj
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ UV460)
Dostawca: AAT BIOQUEST
100410Y0 100410Y1
AATB100410Y0EA 1740 PLN
AATB100410Y0 AATB100410Y1
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ UV460)
Przeciwciała
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Uwaga: For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: mFluor™ UV460
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human