Drukuj

Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (FITC)

Dostawca: AAT BIOQUEST
104411H0 104411H1
AATB104411H0EA 1740 PLN
AATB104411H0 AATB104411H1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (FITC)
Przeciwciała
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: FITC
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR