Drukuj

Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PerCP/Cy5.5)

Dostawca: AAT BIOQUEST
100811U0 100811U1 100811U2
AATB100811U0EA 1500 PLN
AATB100811U0 AATB100811U1 AATB100811U2
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PerCP/Cy5.5)
Przeciwciała
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PerCP/Cy5.5
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR