Drukuj

Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PacBlue)

Dostawca: AAT BIOQUEST
100811J0 100811J1
AATB100811J0EA 1740 PLN
AATB100811J0 AATB100811J1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PacBlue)
Przeciwciała
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PacBlue
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR