Drukuj
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Blue 570)
Dostawca: AAT BIOQUEST
100330T0 100330T1
AATB100330T0EA 1740 PLN
AATB100330T0 AATB100330T1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Blue 570)
Przeciwciała
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Uwaga: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human