Drukuj

Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 514)

Dostawca: AAT BIOQUEST
10033060 10033061
AATB10033060EA 1740 PLN
AATB10033060 AATB10033061
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 514)
Przeciwciała
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 514
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR