Drukuj

Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 514)

Dostawca: AAT BIOQUEST
11610060 11610061
AATB11610060EA 1740 PLN
AATB11610060 AATB11610061
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 514)
Przeciwciała
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 514
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR