Drukuj

Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (PB (Pacific Blue))

Dostawca: AAT BIOQUEST
103401J0 103401J1
AATB103401J0EA 1740 PLN
AATB103401J0 AATB103401J1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (PB (Pacific Blue))
Przeciwciała
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR