Drukuj

Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PB (Pacific Blue))

Dostawca: AAT BIOQUEST
101921J0 101921J1
AATB101921J0EA 1740 PLN
AATB101921J0 AATB101921J1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PB (Pacific Blue))
Przeciwciała
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR