Drukuj

Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Green 620)

Dostawca: AAT BIOQUEST
101610U0 101610U1
AATB101610U0EA 1740 PLN
AATB101610U0 AATB101610U1
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Green 620)
Przeciwciała
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kDa transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: mFluor™ Green 620
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR