Drukuj
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 700)
Dostawca: AAT BIOQUEST
101610J0 101610J1
AATB101610J0EA 1740 PLN
AATB101610J0 AATB101610J1
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 700)
Przeciwciała
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kDa transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Uwaga: For research use only.
Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human