Drukuj

Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 680)

Dostawca: AAT BIOQUEST
100320I0 100320I1
AATB100320I0EA 1740 PLN
AATB100320I0 AATB100320I1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 680)
Przeciwciała
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 680
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR