Drukuj

Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 555)

Dostawca: AAT BIOQUEST
10021090 10021091
AATB10021090EA 1740 PLN
AATB10021090 AATB10021091
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 555)
Przeciwciała
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Uwaga: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 555
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR