Drukuj
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 514)
Dostawca: AAT BIOQUEST
10032060 10032061
AATB10032060EA 1740 PLN
AATB10032060 AATB10032061
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 514)
Przeciwciała
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Uwaga: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 514
Epitope:
Host: Mouse
Isotype:
Reactivity: Human