Drukuj

Anti-BGB-A317 Recombinant Antibody

Dostawca: ProSci
PRSI10-847EA 3150 PLN
PRSI10-847
Anti-BGB-A317 Recombinant Antibody
Przeciwciała
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR