Drukuj

Anti-IgG Goat Polyclonal Antibody (CF®583R)

Dostawca: Biotium
20904-50UL 20904-500UL
20904-50ULEA 523 PLN
20904-50UL 20904-500UL
Anti-IgG Goat Polyclonal Antibody (CF®583R)
Przeciwciała
This is a highly cross-adsorbed goat anti-mouse IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against bovine, horse, human, rabbit, and swine serum. This antibody is not cross-adsorbed against rat. For staining rat tissue, or co-staining with rat antibodies, we recommend using Goat Anti-Mouse IgG (H L), Highly Cross-Adsorbed (Min x Rat).

  • Highly-cross adsorbed for specific staining with minimal background
  • Available with 19 bright and photostable CF® dyes
  • Biotin and HRP conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

CF®583R dyes are spectrally similar red fluorescent dyes with exceptional brightness. These dyes may be excited with a 561 nm to 568 nm laser line and have an emission maximum around 609 nm.

CF®583R is a rhodamine-based fluorescent dye that serves as an alternative to Texas Red®. CF®583R could be used as an excellent energy acceptor in Förster resonance energy transfer (FRET) when paired with R-PE.

CF®583R produce antibody conjugates with brighter fluorescence compared to spectrally similar dyes. In addition, the exceptional water solubility of the dyes facilitate bioconjugation and better preserves biological specificity of the conjugates. CF®583R in amine-reactive forms and with many other bioconjugates.

Abs/Em maxima: 586/609 nm

Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgG (H L)
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR