Drukuj
CELLSTAR® Centrifuge Tubes, Polypropylene, Sterile
Dostawca: Greiner bio-one
CELLSTAR®
GREI188271EA 723 PLN
GREI188271 GREI188281
CELLSTAR® Centrifuge Tubes, Polypropylene, Sterile
Probówki Probówki wirówkowe
These conical-bottom, sterile centrifuge tubes are made from ClearChoice™ polypropylene with excellent optical clarity and high RCF ratings.
  • Sterile
  • Free of RNase, DNase, and human DNA
  • Non-pyrogenic
  • Available with or without a skirt
  • Polypropylene construction
Tubes feature graduations and an ID field printed on the side.
Informacje do zamówienia: Supplied with HDPE flat-top screw caps.