Drukuj
ESD Safety Shoes, Lace-Up, Mattis XXSports Low S1P
Dostawca: ELTEN
720321/35 720321/36 720321/37 720321/38 720321/39 720321/40 720321/41 720321/42 720321/43 720321/44 720321/45 720321/46 720321/47 720321/48
ELTN720321/35EA 507 PLN
ELTN720321/35 ELTN720321/36 ELTN720321/37 ELTN720321/38 ELTN720321/39 ELTN720321/40 ELTN720321/41 ELTN720321/42 ELTN720321/43 ELTN720321/44 ELTN720321/45 ELTN720321/46 ELTN720321/47 ELTN720321/48
ESD Safety Shoes, Lace-Up, Mattis XXSports Low S1P
Obuwie i buty
ESD safety shoes made from textile material with recycled material content and a TPU/PU sole (Wellmaxx Sports). The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides, creating more energy in every step.
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD black
  • Non-metallic, penetration-resistant midsole with recycled material content
  • Steel toe cap
Suitable for use in dry industry, storage, transport, and assembly work areas where there is a risk of penetration from pointed and sharp objects (S1P). Shoes are also suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD) and workplaces on hard undergrounds.
Colour: Black/green
Certyfikaty: EN ISO 20345 S1P SRB, form A.